Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170599.2 | NP_733548.2 | 4 | 696 | ||
GenBank mRNA | AB079880.1 | - | 4 | 533 | ||
AK160146.1 | - | 4 | 659 | |||
BC057555.1 | - | 4 | 696 | 6416190 |