Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085498.2 | NP_001078967.1 | 8 | 1370 | ||
GenBank mRNA | AK172123.1 | - | 6 | 2088 | ||
AK211523.1 | - | 1 | 124 | |||
BC027085.1 | - | 1 | 1485 | |||
BY463843.1 | - | 1 | 245 |