Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085498.2 | NP_001078967.1 | 4, 5 | 415 | ||
GenBank mRNA | AK172123.1 | - | 4, 5 | 382 | ||
AK219882.1 | - | 2, 3 | 143 | |||
BC147754.1 | - | 4, 5 | 410 | 9056703 | ||
BC147755.1 | - | 4, 5 | 410 | 9056704 | ||
BG078687.2 | - | 4, 5 | 330 |