Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145363.2 | NP_663338.2 | 2 | 688 | ||
GenBank mRNA | AK084214.1 | - | 2 | 635 | ||
AK148037.1 | - | 2 | 631 | |||
AK148155.1 | - | 3 | 771 | |||
AK148326.1 | - | 2 | 624 | |||
AK148383.1 | - | 2 | 624 | |||
BC019991.1 | - | 2 | 626 | 4189407 |