Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145369.3 | NP_663344.2 | 2 | 239 | ||
GenBank mRNA | AK075896.1 | - | 2 | 94 | ||
AK131905.1 | - | 2 | 187 | |||
AK131906.1 | - | 2 | 172 | |||
AY669505.1 | - | 2 | 238 | |||
BC019734.1 | - | 2 | 239 | 3987617 |