Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173378.2 | NP_775554.2 | 17, 18 | 3615 | ||
GenBank mRNA | AK133930.1 | - | 2, 3 | 253 | ||
AK134556.1 | - | 10, 11 | 2585 | |||
BC025187.1 | - | 2, 3 | 260 | |||
BC030894.1 | - | 9, 10 | 2244 | |||
BC042874.1 | - | 16, 17 | 3217 |