Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010827.1 | NP_001010827.1 | 1 | 937 |
Gene Symbol: Taar7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010829.1 | NP_001010829.1 | 1 | 937 |
Gene Symbol: Taar7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010835.1 | NP_001010835.1 | 1 | 937 | ||
GenBank mRNA | BC139110.1 | - | 1 | 1003 | 8862132 | |
BC145816.1 | - | 1 | 1003 | 40131119 |
Set Membership: |
G-protein coupled receptor |