Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002602.3 | NP_002593.1 | 2 | 249 | ||
XM_006721942.3 | XP_006722005.1 | 2 | 1222 | |||
XM_017024734.1 | XP_016880223.1 | 2 | 524 | |||
XM_017024735.1 | XP_016880224.1 | 2 | 255 | |||
XM_017024736.1 | XP_016880225.1 | 2 | 189 | |||
GenBank mRNA | BC106884.2 | - | 2 | 206 | 40029486 | |
BQ638010.1 | - | 2 | 190 | |||
BQ639132.1 | - | 2 | 249 | |||
BX537414.1 | - | 2 | 256 | |||
M36476.1 | - | 2 | 206 |