Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172477.4 | NP_766065.1 | 9, 10 | 2263 | ||
GenBank mRNA | AK048041.1 | - | 9, 10 | 2248 | ||
AK076686.1 | - | 1, 2 | 406 | |||
AK081176.1 | - | 9, 10 | 2264 | |||
AK135781.1 | - | 7, 8 | 808 | |||
AK160784.1 | - | 9, 10 | 2025 | |||
AK165202.1 | - | 9, 10 | 2118 | |||
AK167298.1 | - | 9, 10 | 2117 | |||
BC056367.1 | - | 9, 10 | 2133 | |||
BC057310.1 | - | 9, 10 | 2133 | |||
BC145599.1 | - | 8, 9 | 1942 | 9054165 | ||
BC150799.1 | - | 8, 9 | 1942 | 9087710 |