Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iglon5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164518.1 | NP_001157990.1 | 2 | 115 | ||
GenBank mRNA | AK039193.1 | - | 2 | 82 |