Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10222
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
8
601
NM_001001568.1
NP_001001568.1
6
452
NM_001001569.1
NP_001001569.1
5
397
NM_001001570.1
NP_001001570.1
8
622
NM_001001571.1
NP_001001571.1
7
530
NM_001001572.1
NP_001001572.1
5
408
NM_001001573.1
NP_001001573.1
6
588
NM_001001574.1
NP_001001574.1
7
523
NM_001001575.1
NP_001001575.1
4
353
NM_001001576.1
NP_001001576.1
4
312
NM_001001577.1
NP_001001577.1
5
431
NM_001001578.1
NP_001001578.1
6
475
NM_001001579.1
NP_001001579.1
6
468
NM_001001580.1
NP_001001580.1
3
268
NM_001001581.1
NP_001001581.1
7
632
NM_001001582.1
NP_001001582.1
8
710
NM_001001583.1
NP_001001583.1
8
703
NM_001001584.2
NP_001001584.1
7
531
NM_001001585.1
NP_001001585.1
7
655
NM_001315533.1
NP_001302462.1
9
677
NM_002606.2
NP_002597.1
9
781
XM_011529598.2
XP_011527900.1
9
703
XM_011529600.2
XP_011527902.1
9
754
XM_017028366.1
XP_016883855.1
7
519
XM_017028367.1
XP_016883856.1
9
754
GenBank mRNA
AF048837.1
-
9
778
AF067223.1
-
9
730
AF067224.1
-
8
549
AF067225.1
-
6
391
AF067226.1
-
5
336
AK001165.1
-
6
383
AK001307.1
-
8
572
AK294112.1
-
6
418
AK308753.1
-
3
257
AK314679.1
-
9
754
AY196299.1
-
8
595
AY196300.1
-
7
503
AY196301.1
-
5
381
AY196302.1
-
6
561
AY196303.1
-
7
496
AY196304.1
-
4
326
AY196305.1
-
4
285
AY196306.1
-
5
404
AY196307.1
-
6
448
AY196308.1
-
6
441
AY196309.1
-
3
241
AY196310.1
-
7
605
AY196311.1
-
8
683
AY196312.1
-
8
676
AY196313.1
-
7
503
AY196314.1
-
7
628
AY242121.1
-
7
467
AY701187.1
-
9
674
BC009047.1
-
8
575
3874635
BT007016.1
-
8
538
KU178246.1
-
8
538
KU178247.1
-
9
718
KU178248.1
-
5
334
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0