Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173749.4 | NP_776110.3 | 11 | 2018 | ||
GenBank mRNA | AK088017.1 | - | 11 | 1917 | ||
BC031841.1 | - | 11 | 1972 | 3675412 | ||
BC057685.1 | - | 11 | 1972 | 3675412 | ||
CX564812.1 | - | 3 | 672 |