Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181584.4 | NP_853615.1 | 3 | 662 | ||
GenBank mRNA | AK031227.1 | - | 3 | 660 | ||
AY057988.1 | - | 3 | 548 | |||
BC141240.1 | - | 3 | 542 | 9055852 |