Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D15Ertd621e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145959.3 | NP_666071.2 | 17, 18 | 1938 | ||
GenBank mRNA | AK122297.1 | - | 17, 18 | 1788 | ||
AK137324.1 | - | 17, 18 | 1939 | |||
BC033609.1 | - | 9, 10 | 1020 | 5401763 | ||
BC070426.1 | - | 17, 18 | 1924 | 6405871 | ||
DQ228142.1 | - | 17, 18 | 1717 |