Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033242.1 | NP_001028414.1 | 4 | 884 | ||
GenBank mRNA | AK030852.1 | - | 1 | 1745 | ||
AK032293.1 | - | 4 | 699 | |||
AK043247.1 | - | 4 | 808 | |||
AK085741.1 | - | 4 | 885 | |||
AK144635.1 | - | 4 | 775 | |||
AK207214.1 | - | 1 | 142 | |||
AK207852.1 | - | 1 | 135 | |||
AK208795.1 | - | 1 | 135 | |||
BC025487.1 | - | 4 | 741 | |||
BC141314.1 | - | 4 | 778 | 9055926 | ||
BC141315.1 | - | 4 | 778 | 9055927 |