Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145378.4 | NP_663353.3 | 20 | 2345 | ||
GenBank mRNA | AK037218.1 | - | 17 | 3794 | ||
AK137721.1 | - | 19 | 2574 | |||
BC042758.1 | - | 8 | 1224 | |||
DQ888308.1 | - | 19 | 2253 |
Gene Symbol: Gm28042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB195274.1 | - | 27 | 3578 | ||
AB195275.1 | - | 26 | 5686 | |||
BC098210.1 | - | 21 | 2739 |