Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127363.1 | NP_001120835.1 | 6, 7 | 751 | ||
NM_183144.3 | NP_898967.2 | 6, 7 | 751 | |||
GenBank mRNA | AK083449.1 | - | 7, 8 | 750 | ||
AK157558.1 | - | 6, 7 | 614 | |||
AK171360.1 | - | 6, 7 | 751 | |||
AK171880.1 | - | 6, 7 | 727 | |||
BC056341.1 | - | 6, 7 | 751 | 5703345 |