Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162941.1 | NP_001156413.1 | 4 | 378 | ||
NM_001162942.1 | NP_001156414.1 | 4 | 321 | |||
NM_153058.4 | NP_694698.3 | 3 | 429 | |||
GenBank mRNA | AI325879.1 | - | 4 | 315 | ||
AK028823.1 | - | 4 | 379 | |||
AK038562.1 | - | 3 | 416 | |||
AK040767.1 | - | 3 | 404 | |||
AK045493.1 | - | 3 | 312 | |||
AK045904.1 | - | 3 | 404 | |||
AK046095.1 | - | 3 | 371 | |||
AK046221.1 | - | 3 | 396 | |||
AK148954.1 | - | 3 | 404 | |||
AK168834.1 | - | 5 | 263 | |||
AK188282.1 | - | 118 | ||||
AK192344.1 | - | 1 | 13 | |||
BC025804.1 | - | 3 | 376 | 5101879 | ||
BC027056.1 | - | 3 | 395 | 5361238 | ||
BC028987.1 | - | 3 | 351 | 5362960 | ||
BC035254.1 | - | 3 | 381 | 5363176 |