Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033531.3 | NP_001028703.2 | 4 | 710 | ||
NM_001163020.1 | NP_001156492.1 | 4 | 710 | |||
GenBank mRNA | AK122569.1 | - | 333 | |||
AK134944.1 | - | 4 | 682 | |||
BC141239.1 | - | 3 | 315 | 9055851 | ||
CA556185.2 | - | 3 | 302 |