Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145384.2 | NP_663359.2 | 7, 8 | 892 | ||
GenBank mRNA | AK081661.1 | - | 7, 8 | 879 | ||
AK156668.1 | - | 7, 8 | 892 | |||
BC019216.1 | - | 7, 8 | 863 | 5065163 | ||
BC025609.1 | - | 6, 7 | 830 | 5309871 |