Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177628.2 | NP_001171099.1 | 3, 4 | 1314 | ||
NM_145977.2 | NP_666089.1 | 3, 4 | 1395 | |||
GenBank mRNA | AK035428.1 | - | 3, 4 | 1264 | ||
AK135655.1 | - | 3, 4 | 586 | |||
BC031381.1 | - | 3, 4 | 1395 | 5050610 | ||
BC034084.1 | - | 1, 2 | 644 |