Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172515.2 | NP_766103.2 | 12 | 1310 | ||
GenBank mRNA | AK029899.1 | - | 12 | 1303 | ||
AK133102.1 | - | 12 | 1361 | |||
BC050789.1 | - | 12 | 1310 | 6706914 | ||
BC100435.1 | - | 12 | 1315 |