Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253719.1 | NP_001240648.1 | 9 | 1311 | ||
NM_178673.4 | NP_848788.2 | 9 | 1571 | |||
GenBank mRNA | AF374460.1 | - | 9 | 1545 | ||
AK033652.1 | - | 9 | 1562 | |||
AK046070.1 | - | 9 | 1312 | |||
AK047518.1 | - | 1 | 2194 | |||
AK081152.1 | - | 9 | 1562 | |||
AK138388.1 | - | 9 | 1863 | |||
BC075640.1 | - | 9 | 1534 | 30545529 |