Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025608.2 | NP_001020779.1 | 4 | 869 | ||
NM_001277195.1 | NP_001264124.1 | 4 | 866 | |||
GenBank mRNA | AK027934.1 | - | 3 | 782 | ||
AK145912.1 | - | 3 | 789 | |||
AK171420.1 | - | 3 | 779 | |||
AK171702.1 | - | 3 | 794 | |||
AK208706.1 | - | 1 | 78 | |||
BC066992.1 | - | 4 | 817 | 5685985 |