Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033253.3 | NP_001028425.1 | 2 | 552 | ||
NM_001159942.1 | NP_001153414.1 | 3 | 718 | |||
GenBank mRNA | AK033676.1 | - | 3 | 718 | ||
AK034403.1 | - | 3 | 719 | |||
AK045140.1 | - | 3 | 898 | |||
AK135923.1 | - | 2 | 554 | |||
BC117965.1 | - | 1 | 154 | 40098326 | ||
BC117966.1 | - | 1 | 147 | 40098330 |