Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145985.4 | NP_666097.3 | 8 | 1323 | ||
GenBank mRNA | AK028556.1 | - | 8 | 1328 | ||
AK047995.1 | - | 8 | 1363 | |||
AK128973.1 | - | 8 | 1328 | |||
AK147747.1 | - | 8 | 1357 | |||
AK154066.1 | - | 1327 | ||||
AK170005.1 | - | 4 | 510 | |||
AK204777.1 | - | 3 | 255 | |||
BC017124.1 | - | 8 | 1318 | 4480874 | ||
BC023728.1 | - | 8 | 1342 | 5351777 | ||
BC024127.1 | - | 7 | 943 | |||
BC033387.1 | - | 8 | 1323 | 5368060 | ||
BC034754.1 | - | 8 | 1300 | 4954556 | ||
BC050874.1 | - | 1 | 130 | |||
BC083152.1 | - | 1316 | 30461105 |