Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 5 | 941 | ||
NM_001282677.1 | NP_001269606.1 | 5 | 941 | |||
NM_001282678.1 | NP_001269607.1 | 5 | 976 | |||
GenBank mRNA | AB008112.1 | - | 5 | 912 | ||
AB052090.1 | - | 5 | 845 | |||
AB052091.1 | - | 5 | 845 | |||
AB052092.1 | - | 5 | 845 | |||
AB052093.1 | - | 5 | 845 | |||
AB052094.1 | - | 5 | 845 | |||
AF026086.1 | - | 5 | 905 | |||
AF030356.1 | - | 5 | 849 | |||
AK292955.1 | - | 5 | 846 | |||
AK293757.1 | - | 5 | 884 | |||
BC035575.1 | - | 5 | 883 | 5498274 |