Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000466.2 | NP_000457.1 | 4 | 535 | ||
NM_001282677.1 | NP_001269606.1 | 4 | 535 | |||
NM_001282678.1 | NP_001269607.1 | 4 | 570 | |||
GenBank mRNA | AB008112.1 | - | 4 | 506 | ||
AB052090.1 | - | 4 | 439 | |||
AB052091.1 | - | 4 | 439 | |||
AB052092.1 | - | 4 | 439 | |||
AB052093.1 | - | 4 | 439 | |||
AB052094.1 | - | 4 | 439 | |||
AF026086.1 | - | 4 | 499 | |||
AF030356.1 | - | 4 | 443 | |||
AK292955.1 | - | 4 | 440 | |||
AK293757.1 | - | 4 | 478 | |||
BC035575.1 | - | 4 | 477 | 5498274 | ||
DA313282.1 | - | 4 | 536 | |||
DC370317.1 | - | 4 | 546 |