Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256104.1 | NP_001243033.1 | 2 | 1031 | ||
NM_145406.2 | NP_663381.1 | 3 | 1100 | |||
GenBank mRNA | AK041958.1 | - | 3 | 1051 | ||
BC023050.1 | - | 2 | 976 | 5355849 | ||
BC027440.1 | - | 3 | 1007 | 3980056 |