Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chtf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145412.3 | NP_663387.3 | 3 | 254 | ||
GenBank mRNA | AK030924.1 | - | 3 | 253 | ||
AK088257.1 | - | 3 | 263 | |||
AK165622.1 | - | 3 | 236 | |||
AK184621.1 | - | 2 | 172 | |||
BC023107.1 | - | 1 | 26 | 3966700 |