Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144818.3 | NP_659067.2 | 2 | 314 | ||
GenBank mRNA | AK028919.1 | - | 2 | 299 | ||
AK042706.1 | - | 2 | 231 | |||
AK080433.1 | - | 2 | 303 | |||
BC021499.1 | - | 2 | 288 | 5346298 | ||
BC025060.1 | - | 2 | 239 | 3673890 | ||
BY772780.1 | - | 2 | 316 |