Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145743.2 | NP_665686.2 | 11 | 1338 | ||
GenBank mRNA | AF397909.1 | - | 10 | 1041 | ||
AF520417.1 | - | 11 | 1187 | |||
AK044595.1 | - | 11 | 1214 | |||
AK170996.1 | - | 12 | 1381 | |||
BC023308.1 | - | 10 | 1015 | |||
BC089595.1 | - | 11 | 1188 | 30118411 |