Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003950.2 | NP_001003950.1 | 3 | 624 | ||
GenBank mRNA | AK152122.1 | - | 3 | 676 | ||
AK178592.1 | - | 1 | 184 | |||
AK203553.1 | - | 1 | 184 | |||
BC080289.1 | - | 3 | 624 | 6816077 |