Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090155.1 | - | 1608 |
Gene Symbol: Lgalsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173752.4 | NP_776113.1 | 4 | 398 | ||
GenBank mRNA | AK041053.1 | - | 4 | 313 | ||
AK081513.1 | - | 1 | 83 | |||
AK156350.1 | - | 4 | 526 | |||
AK167557.1 | - | 4 | 336 | |||
BC019131.1 | - | 4 | 369 | 5036749 |