Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175002.2 | NP_778167.1 | 2 | 533 | ||
GenBank mRNA | AK004099.1 | - | 2 | 370 | ||
AK131977.1 | - | 2 | 423 | |||
BC025076.1 | - | 1 | 304 | 4459181 | ||
BC096583.1 | - | 2 | 477 | 5025927 | ||
EU069462.1 | - | 1 | 191 |