Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017426.1 | NP_001017426.1 | 17 | 4536 | ||
GenBank mRNA | AK129121.1 | - | 7 | 1328 | ||
AK136019.1 | - | 7 | 717 | |||
BC038313.1 | - | 2 | 174 | |||
BC075632.1 | - | 18 | 4712 | 30362899 |