Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144824.2 | NP_659073.1 | 9 | 1530 | ||
GenBank mRNA | AK077570.1 | - | 9 | 1434 | ||
AK145219.1 | - | 9 | 1457 | |||
AK169271.1 | - | 8 | 1543 | |||
BC021790.1 | - | 9 | 1530 | 4220923 | ||
BC069868.1 | - | 10 | 1574 | 5101220 |