Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178367.4 | NP_848144.3 | 4 | 733 | ||
GenBank mRNA | AK035177.1 | - | 4 | 733 | ||
AK087592.1 | - | 2 | 2489 | |||
BC027842.1 | - | 4 | 730 | |||
BC052172.1 | - | 4 | 727 | 6515901 |