Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004144.1 | NP_001004144.1 | 18, 19 | 2216 | ||
GenBank mRNA | AK050701.1 | - | 5, 6 | 534 | ||
BC079870.1 | - | 18, 19 | 2216 | 6831910 |