siRNA Details
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145431.2 | NP_663406.2 | 4, 5 | 491 | ||
GenBank mRNA | AK141230.1 | - | 4, 5 | 466 | ||
AK170853.1 | - | 4, 5 | 464 | |||
BC018399.1 | - | 4, 5 | 482 | 3491925 | ||
BC035421.1 | - | 4, 5 | 482 |

More Information
Other Species Targeted

