Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098836.1 | NP_001092306.1 | 8, 9 | 853 | ||
NM_001098837.1 | NP_001092307.1 | 8, 9 | 832 | |||
GenBank mRNA | AK090368.1 | - | 8, 9 | 834 | ||
BC059880.1 | - | 2, 3 | 248 | |||
BC130269.1 | - | 7, 8 | 603 | |||
BM950535.1 | - | 6, 7 | 410 |