Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015099.2 | NP_001015099.2 | 5 | 510 | ||
NM_001167963.1 | NP_001161435.1 | 5 | 510 | |||
NM_001167964.1 | NP_001161436.1 | 5 | 510 | |||
GenBank mRNA | AK031333.1 | - | 5 | 370 | ||
AK033513.1 | - | 5 | 452 | |||
AK084256.1 | - | 5 | 381 | |||
AK129332.1 | - | 5 | 413 | |||
BC086455.1 | - | 5 | 383 | 30458160 |