Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146035.2 | NP_666147.1 | 1 | 1699 | ||
GenBank mRNA | AK083370.1 | - | 1 | 1656 | ||
AK152925.1 | - | 1 | 1668 | |||
AK159185.1 | - | 2 | 1573 | |||
AK166037.1 | - | 1 | 1664 | |||
AK171000.1 | - | 1 | 1702 | |||
BC010583.1 | - | 1 | 1644 | 3500709 | ||
BC027169.1 | - | 1 | 1644 | 4954047 |