Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175644.3 | NP_783575.2 | 3 | 435 | ||
GenBank mRNA | AF467771.1 | - | 2 | 321 | ||
AK078727.1 | - | 3 | 437 | |||
BC111115.1 | - | 2 | 276 | |||
BC139462.1 | - | 3 | 375 | 8862484 | ||
BC139463.1 | - | 3 | 375 | 8862485 |