Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tppp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128634.1 | NP_001122106.1 | 3 | 476 | ||
GenBank mRNA | BC049709.1 | - | 4 | 590 | 6740792 |