Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177338.5 | NP_796312.2 | 7 | 1247 | ||
GenBank mRNA | AK052729.1 | - | 7 | 1257 | ||
AK089782.1 | - | 7 | 1248 | |||
BC002212.1 | - | 5 | 651 | |||
BC051457.1 | - | 7 | 1250 | 6398463 |