Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013753.2 | NP_001013775.2 | 1 | 4142 | ||
GenBank mRNA | AK139275.1 | - | 1 | 2403 | ||
AK142358.1 | - | 1 | 4138 | |||
AK147265.1 | - | 1 | 4144 | |||
AK158668.1 | - | 1 | 1530 | |||
BC079850.1 | - | 1 | 2139 | |||
BC089004.1 | - | 1 | 2031 | 6841033 |