Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081302.1 | NP_001074771.1 | 57 | 9616 | ||
GenBank mRNA | AK153924.1 | - | 10 | 1706 | ||
BC035955.1 | - | 8 | 1253 | |||
BC051169.1 | - | 16 | 3083 |