Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205345.1 | NP_001192274.1 | 15 | 1967 | ||
NM_001205346.1 | NP_001192275.1 | 15 | 1961 | |||
NM_194342.3 | NP_919323.2 | 14 | 1871 | |||
GenBank mRNA | AK047688.1 | - | 3 | 314 | ||
AK128958.1 | - | 14 | 1865 | |||
AK156246.1 | - | 15 | 1968 | |||
AK172398.1 | - | 15 | 1969 | |||
AK172996.1 | - | 13 | 2571 | |||
AY682987.1 | - | 14 | 1727 | |||
BC023965.1 | - | 4 | 519 | |||
BC098208.1 | - | 14 | 2212 | 6827666 | ||
BQ879674.1 | - | 2 | 83 |