Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145477.2 | NP_663452.1 | 5, 6 | 782 | ||
GenBank mRNA | BC021379.1 | - | 5, 6 | 755 | 5044319 |